Anti-CD6 Monoclonal Antibody [clone: 3F7B6]

Supplier: Adipogen
ANC-151-820
102511-932EA 297.69 USD
102511-932
Anti-CD6 Monoclonal Antibody [clone: 3F7B6]
Antibodies
Human CD6 is expressed on peripheral blood T cells, most thymocytes and a subset of B cells. CD6 is similar to CD5 and contains three cysteine-rich domains with structural homology to the macrophage scavenger receptor. CD6 is involved in T cell activation and its ligand was recently identified.

  • preservative free

Type: Primary
Antigen: CD6
Clonality: Monoclonal
Clone: 3F7B6
Conjugation: Preservative Free
Epitope:
Host:
Isotype: Mouse IgG1kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR