Anti-CD4 Monoclonal Antibody [clone: M-T441]

Supplier: Adipogen
ANC-148-820
102511-910EA 297.69 USD
102511-910
Anti-CD4 Monoclonal Antibody [clone: M-T441]
Antibodies
Human CD4 is a cell surface glycoprotein of 55kDa expressed on most thymocytes, on about two thirds of peripheral T cells and on some monocyte macrophage lineage cells. CD4 interacts with MHC Class II molecules in an accessory role during foreign antigen recognition by T cells. The extracellular portion of CD4 is an array of four Ig-like domains. The N-terminal region of CD4 is a receptor for the HIV-1 viral protein gp120.

  • preservative free

Type: Primary
Antigen: CD4
Clonality: Monoclonal
Clone: M-T441
Conjugation: Preservative Free
Epitope:
Host:
Isotype: Mouse IgG2b
Reactivity: Baboon, Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR