Anti-CD1a Monoclonal Antibody (R-PE) [clone: CB-T6]

Supplier: Adipogen
ANC-145-050
102511-880EA 447.23 USD
102511-880
Anti-CD1a Monoclonal Antibody (R-PE) [clone: CB-T6]
Antibodies
The human CD1 genetic locus consists of five closely related genes. Three of the gene products (CD1a, CD1b and CD1c) have been officially clustered using antibodies and are expressed on cortical thymocytes, a subset of B cells, and on most professional antigen presenting cells. CD1a+/CD11c+ dendritic cells are direct precursors of Langerhans cells in peripheral blood. CD1 molecules have a specialized lipid antigen-presenting function.

Type: Primary
Antigen: CD1a
Clonality: Monoclonal
Clone: CB-T6
Conjugation: R-PE
Epitope:
Host:
Isotype: Mouse IgG1kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR