Anti-IgM Monoclonal Antibody [clone: UCHB1]

Supplier: Adipogen
ANC-141-820
102511-828EA 297.69 USD
102511-828
Anti-IgM Monoclonal Antibody [clone: UCHB1]
Antibodies
Human surface membrane bound IgM complexes with a heterodimer of transmembrane proteins of Igalpha and Igbeta (CD79a and CD79b) to form the B cell antigen receptor (BCR). The BCR complex plays an important role in B cell development and activation. The transmembrane region of mIgM is essential for functional BCR complex assembly.

  • preservative free

Type: Secondary
Antigen: IgM
Clonality: Monoclonal
Clone: UCHB1
Conjugation: Preservative Free
Epitope:
Host:
Isotype: Mouse IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR