Anti-IgG Monoclonal Antibody (Biotin) [clone: ICO-97]

Supplier: Adipogen
ANC-139-030
102511-820EA 350.31 USD
102511-820
Anti-IgG Monoclonal Antibody (Biotin) [clone: ICO-97]
Antibodies
Human immunoglobulins are glycoproteins composed of two disulfide-bonded heavy (H) chain subunits, each of which is linked by interchain disulfide bonds to a light (L) chain forming a tetramolecular complex. There are five classes of immunoglobulins, designated IgG, IgA, IgM, IgD and IgE, which are defined by differences in the constant region of H chains. L chains are divided into kappa or lambda classifications based on structural antigenic differences. All classes of immunoglobulins have been found on the cell surface of B lymphocytes where they function as antigen receptors to elicit antigen-dependent proliferation and secretion of antigen specific soluble circulating antibodies.

Type: Secondary
Antigen: IgG
Clonality: Monoclonal
Clone: ICO-97
Conjugation: Biotin
Epitope:
Host:
Isotype: Mouse IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR