Anti-IgA Monoclonal Antibody [clone: Hisa43]

Supplier: Adipogen
ANC-137-020
102511-796EA 297.69 USD
102511-796
Anti-IgA Monoclonal Antibody [clone: Hisa43]
Antibodies
Human immunoglobulins are glycoproteins composed of two disulfide-bonded heavy (H) chain subunits, each of which is linked by interchain disulfide bonds to a light (L) chain forming a tetramolecular complex. There are five classes of immunoglobulins, designated IgG, IgA, IgM, IgD and IgE, which are defined by differences in the constant region of H chains. L chains are divided into kappa or lambda classifications based on structural antigenic differences. All classes of immunoglobulins have been found on the cell surface of B lymphocytes where they function as antigen receptors to elicit antigen-dependent proliferation and secretion of antigen specific soluble circulating antibodies.

Type: Secondary
Antigen: IgA
Clonality: Monoclonal
Clone: Hisa43
Conjugation: Unconjugated
Epitope:
Host:
Isotype: Mouse IgG1kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR