Anti-MHC1 Monoclonal Antibody (Biotin) [clone: 3F10]

Supplier: Adipogen
ANC-121-030
102511-760EA 350.31 USD
102511-760
Anti-MHC1 Monoclonal Antibody (Biotin) [clone: 3F10]
Antibodies
Human MHC Class I (HLA-A, B & C) molecules are expressed on nearly all nucleated cells. MHC Class I molecules consist of heterodimers of a highly polymorphic transmembrane alpha chain and an invariant beta chain. MHC Class I molecules bind to endogenously synthesized peptides of about 8-10 amino acids which are presented to CD8+ T cells. Evidence indicates exogenously derived peptides are also presented by MHC Class I molecules.

Type: Primary
Antigen: MHC1
Clonality: Monoclonal
Clone: 3F10
Conjugation: Biotin
Epitope:
Host:
Isotype: Mouse IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR