Anti-CD80 Monoclonal Antibody [clone: P1.H1.A1.A1]

Supplier: Adipogen
ANC-110-020
102511-748EA 350.31 USD
102511-748
Anti-CD80 Monoclonal Antibody [clone: P1.H1.A1.A1]
Antibodies
Human CD80 (B7-1) is a costimulatory ligand for CD28 and CD152 (CTLA-4). CD80 is expressed on activated B cells.

Type: Primary
Antigen: CD80
Clonality: Monoclonal
Clone: P1.H1.A1.A1
Conjugation: Unconjugated
Epitope:
Host:
Isotype: Mouse IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR