Anti-TCR Monoclonal Antibody (R-PE) [clone: Jovi-3]

Supplier: Adipogen
ANC-102-050
102511-746EA 670.15 USD
102511-746
Anti-TCR Monoclonal Antibody (R-PE) [clone: Jovi-3]
Antibodies
Human T cell receptor (TCR) is a T cell surface expressed heterodimer of alpha/beta or gamma/delta chains. TCR’s recognize antigen peptides bound to MHC molecules forming the basis of antigen specific response by T cells.

Type: Primary
Antigen: TCR
Clonality: Monoclonal
Clone: Jovi-3
Conjugation: R-PE
Epitope:
Host:
Isotype: Mouse IgG2alambda
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR