Anti-TCR Monoclonal Antibody [clone: Jovi-3]

Supplier: Adipogen
ANC-102-020
102511-738EA 372.46 USD
102511-738
Anti-TCR Monoclonal Antibody [clone: Jovi-3]
Antibodies
Human T cell receptor (TCR) is a T cell surface expressed heterodimer of alpha/beta or gamma/delta chains. TCR’s recognize antigen peptides bound to MHC molecules forming the basis of antigen specific response by T cells.

Type: Primary
Antigen: TCR
Clonality: Monoclonal
Clone: Jovi-3
Conjugation: Unconjugated
Epitope:
Host:
Isotype: Mouse IgG2alambda
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR