Anti-Rab6-GTP Human Antibody (ATTO 488) [clone: AA2]

Supplier: Adipogen
AG-27B-0004TD-C100
102511-420EA 526.15 USD
102511-420
Anti-Rab6-GTP Human Antibody (ATTO 488) [clone: AA2]
Antibodies
Rab6 is involved in protein transport. It is a regulator of membrane traffic from the Golgi apparatus towards the endoplasmic reticulum (ER).

  • Purified from HEK 293 cell culture supernatant.

Keep conjugated formats at +4°C. Stable for at least 1 year after receipt when stored at +4°C.

Type:
Antigen: Rab6-GTP
Clonality:
Clone: AA2
Conjugation: ATTO 488
Epitope:
Host: Human
Isotype: IgG2 lambda
Reactivity: Drosophila, Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR