Anti-Neutrophils Rat Monoclonal Antibody [clone: Nimp-R14]

Supplier: Adipogen
AG-20B-0043PF-C500 AG-20B-0043PF-M002
102511-412EA 387.69 USD
102511-412 102511-414
Anti-Neutrophils Rat Monoclonal Antibody [clone: Nimp-R14]
Antibodies
Neutrophils are key components of the inflammatory response and as such contribute to the killing of microorganisms. In addition, recent evidence suggests their involvement in the development of the immune response. The Nimp-R14 mAb displays features of an optimal reagent allowing the selective depletion of mouse neutrophils in vivo.

  • Purified from concentrated hybridoma tissue culture supernatant.
  • Purity: >95% (SDS-PAGE)

Stable for at least 1 year after receipt when stored at -20°C.
Preservative-free

Type:
Antigen: Neutrophils
Clonality: Monoclonal
Clone: Nimp-R14
Conjugation:
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR