Anti-BAFF-R Rat Monoclonal Antibody (ATTO 647N) [clone: 9B9]

Supplier: Adipogen
AG-20B-0034TS-T100
102511-404EA 526.15 USD
102511-404
Anti-BAFF-R Rat Monoclonal Antibody (ATTO 647N) [clone: 9B9]
Antibodies
Three receptors for BAFF were recently identified and designated TACI, BCMA and BAFF-R. BAFF-R signaling is essential for B cell development, survival and humoral immune responses.

  • Purified from concentrated hybridoma tissue culture supernatant.
  • Purity: >95% (SDS-PAGE)

Keep conjugated formats at +4°C.

Type:
Antigen: BAFF-R
Clonality: Monoclonal
Clone: 9B9
Conjugation: ATTO 647N
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR