Anti-TRAIL-R1 Mouse Monoclonal Antibody [clone: HS101]

Supplier: Adipogen
AG-20B-0022PF-C100
102511-390EA 512.31 USD
102511-390
Anti-TRAIL-R1 Mouse Monoclonal Antibody [clone: HS101]
Antibodies
TRAIL-R1 is a receptor for the cytotoxic ligand TRAIL. The adapter molecule FADD recruits caspase-8 to the activated receptor. The resulting death-inducing signaling complex (DISC) performs caspase-8 proteolytic activation which initiates the subsequent cascade of caspases (aspartate-specific cysteine proteases) mediating apoptosis. Promotes the activation of NF-kappaB.

  • Stable for at least 1 year after receipt when stored at -20°C.

Purified from concentrated hybridoma tissue culture supernatant.

Purity: >95% (SDS-PAGE)

Type:
Antigen: TRAIL-R1
Clonality: Monoclonal
Clone: HS101
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR