NEBNEXT® Fast DNA Fragmentation and Library Prep Set for Ion Torrent, New England Biolabs
Supplier: New England Biolabs (NEB)
E6285L
102500-086EA 1252 USD
102500-086
NEBNEXT® Fast DNA Fragmentation and Library Prep Set for Ion Torrent, New England Biolabs
Nucleic Acid Reagents Next Generation Sequencing Reagents
The NEBNext® Fast DNA Fragmentation & Library Prep Set for Ion Torrent™ contains enzymes and buffers in convenient master mix formulations that are ideally suited for sample preparation for next-generation sequencing on the Ion Torrent Sequencer (Life Technologies, Inc.)
Each of these components must pass rigorous quality control standards and are lot controlled, both individually and as a set of reagents.