StabilGuard® Choice Stabilizer, Surmodics
Supplier: Surmodics
StabilGuard® Choice Stabilizer protects the activity of proteins while simultaneously blocking nonspecific binding sites on microspheres. In microarray applications, the conformation and activity of proteins are effectively preserved in a dried state when stored with StabilGuard Choice stabilizer. The product ultimately increases signal and reduces background to improve the sensitivity of assays.
- Protein-Free
- 3 year shelf life
- Increased stability
- Reduces non-specific binding
StabilGuard Choice Microarray Stabilizer (BSA-Free) improves the stability of proteins covalently bound to microspheres stored in solution and array proteins stored in the dried state, without the use of BSA protein.
Certifications: ISO 13485:2016 & 9001:2015 Certified