Mini-Run Electrophoresis System, Bulldog Bio
Supplier: Bulldog Bio
BMR100
102408-078EA 761.54 USD
102408-078
Mini-Run Electrophoresis System, Bulldog Bio
Electrophoresis Systems Horizontal Gel Electrophoresis Systems
Down size and pay less.
The Mini-Run horizontal gel apparatus can be used for almost any agarose gel electrophoresis application. The chamber has a built-in DC power supply so you can just plug it in and be ready to run your gel.
The cover and gel chamber are both made from an advanced etch-resistant plastic and the electrodes are platinum coated for clean operation and durability.