Nippon Genetics Bambanker DIRECT Cell Freezing Medium for Hybridomas, Bulldog Bio
Supplier: Bulldog Bio
BBD01
102407-916EA 162.11 USD
102407-916
Nippon Genetics Bambanker DIRECT Cell Freezing Medium for Hybridomas, Bulldog Bio
Cell Freezing Media
Long-Term Storage of Hybridomas at -80 °C or in Liquid Nitrogen.
  • Up to 3× Better Recovery Rates
  • No Step-wise Freezing
There are certain cell types, such as hybridomas, which exhibit increased sensitivity to “over” manipulations. These cells often exhibit higher death rates or unwanted differentiation after long-term term storage and freeze thaw cycling.
It is for these types of cells that Bambanker DIRECT was created – to provide tight environmental control. By eliminating the centrifugation step, cells can now be frozen for long-term storage immediately upon addition of the novel cryoprotectant.
Just add Bambanker DIRECT, one-to-one, with your cells in media and directly place in freezer. And like with the original Bambanker, the DIRECT version has no serum, so it is great for cells which animal-derived serum may be an issue.