AcquaStain Protein Gel Stain, Bulldog Bio
Supplier: Bulldog Bio
AS001000
102407-910EA 150.08 USD
102407-910
AcquaStain Protein Gel Stain, Bulldog Bio
Stains Electrophoresis Stains
The best Coomassie Blue stain on the market; one step, ten minutes, zero destaining, and zero toxicity.
  • One-step staining, ideal for high throughput and teaching labs
  • Bands appear in seconds, but never over-saturate
  • No additional equipment, prewashing, destaining, or gel wrinkling
  • Safe for students and the environment
  • Reusable for extra savings
AcquaStain is a single-step protein gel dye. This novel product requires no pre-or post-treatment of the gel, requires about ten minutes to use for most applications, and yields sharp bands in seconds. AcquaStain is water-based and contains no harmful chemicals or acids, so it is much safer than standard dyes. It also causes no observable gel wrinkling or shrinkage, and can even be disposed of down a sink after staining. Better still, and unlike methanol-based stains, protein sensitivity approaches that of silver staining techniques. It can even be used for mass spectrometry.