FastGene Agarose Tablets, Bulldog Bio
Supplier: Bulldog Bio
AG05
102407-908PK 221.92 USD
102407-908
FastGene Agarose Tablets, Bulldog Bio
Electrophoresis Gels Agarose and Acrylamide
No Weighing Required.
Just add one tablet to 50 ml of gel running buffer and heat—the result is a 1% agarose gel. It’s that simple. With our FastGene Agarose Tablets there’s no powder to weigh and no mess to make!