BioShake iQ Thermomixer, Bulldog Bio
Supplier: Bulldog Bio
1808-0506
102407-834EA 3748.46 USD
102407-834
BioShake iQ Thermomixer, Bulldog Bio
Block Heaters Shaking Block Heaters
For precise control of speed and temperature in a small package, the BioShake iQ has no equal. Boasting the same established mixing mechanics utilized in all BioShake units, the BioShake iQ adds the feature of full thermal control over the shaking samples. The unit is housed in a conveniently small footprint, no bigger than two microplates placed side by side, and delivers well-blended samples in a variety of vessels including microplates, tubes, and vials.
The BioShake iQ Thermomixer offers an ultra-efficient, totally controlled shaking axis to ensure samples mix completely and quickly
To ensure whisper quiet operation, the device is designed to shake vessels only and thoroughly mix all samples, without vibrating an entire lab bench or robotic deck. A patented dampening system relies upon counter-weight symmetry to ensure noticeable noise reduction in any work environment.
The orbital shaking is so precisely controlled, it eliminates the need to shake down samples after mixing. Even high density plates or low sample volumes offer no obstacle for these precision units.
Ordering information: For heating capability, the BioShake iQ must be ordered with an adaptor (such as the 1808-1021 flat bottom plate adaptor). The BioShake iQ Thermomixer is supplied with a two-year parts and labor warranty.