BioShake XP Tube and Plate Mixer, Bulldog Bio
Supplier: Bulldog Bio
1808-0505
102407-832EA 3437.08 USD
102407-832
BioShake XP Tube and Plate Mixer, Bulldog Bio
Shakers and Mixers Microplate Shakers
BioShake Plate mixers offer an ultra-efficient, totally controlled shaking axis so that samples mix completely in a fraction of the time of competing systems.
  • Shaking speed from 200-3000 rpm
  • The orbital shaking is so precisely controlled, so need to spin down the plates after mixing
  • Even high density plates, or low sample volumes, offer no obstacle for these precision tools
  • It guarantees fast, splatter-free, mixing for tubes, glass vials or across an entire 1536-well microplate