Transofix™ Transfer Spike, B.Braun
Supplier: B Braun
415125
10222-534CS 218.28 USD
10222-534
Transofix™ Transfer Spike, B.Braun
Infusions
This features a double spike for rapid transfer of medication or diluent between rubber stoppered containers.
  • Sterile
  • Total shelf life of 60 months
  • Ethylene oxide sterlization method
Components are not made with DEHP, BPA, Natural rubber or PVC.
Certifications: CE marked.