Free ATP/Total, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS
AQ100X
10222-312PK 461.5 USD
10222-312 10222-314
Free ATP/Total, Hardy Diagnostics
Media for Microbiology Additives and Supplements for Microbiology
Aquasnap™ Free ATP, for use with Hygiena luminometers
  • All-in one testing device
  • Simple, quick, and easy to use
  • Results in seconds
  • Unique liquid-stable reagent provides unprecedented accuracy and reproducibility
  • 12-month shelf life at a refrigerated temperature of 2-8°C
  • 4-week shelf life at room temperature, 21°C
  • Tolerant to temperature flucuations
  • Collects 100ul of sample
  • Costs up to 40% less than other ATP water tests
AquaSnap Free detects non-microbial (free) ATP in solution
AquaSnap Total detects both microbial and non-microbial (free) ATP in solution. Used alongside AquaSnap Free, the difference between AquaSnap Total and AquaSnap Free provides an estimation of microbial ATP in a sample.
AquaSnap contains liquid-stable chemistry in the patented Snap-Valve™ bulb. This unique chemistry and design eliminates the need to reconsititute lyophilized pellets, enabling consistent results. The chemistry in AquaSnap is a high power, ultra-sensitive formulation to deliver precise results for low levels of ATP, in water.
Used alongside AquaSnap Total, the difference between AquaSnap Total and AquaSnap Free provides an estimation of microbial ATP in a sample. Aquasnap™ is a user-friendly and self contained ATP water testing device used in conjunction with Hygiena luminometers. The pen-sized testing device is easy to use, economical, and gives real-time results. The honeycomb-shaped dipper collects 100ul of water for consistent sample collection. It uses a liquid-stable reagent instead of freeze-dried enzymes, giving it superior reproducibility and sensitivity.