Methyl red reagent

Supplier: HARDY DIAGNOSTICS

Z117
10222-176EA 60.89 USD
10222-176
Methyl red reagent
Methyl red

Developer reagent for the Methyl Red test.


The Hardy Diagnostics manufacturing facility and quality management system comply with the FDA's Good Manufacturing Practices (GMP) and have achieved ISO 13485 certification as a medical device manufacturer.

Formula: C₁₅H₁₅N₃O₂
MDL Number: MFCD00002425
CAS Number: 493-52-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR