Cellvation Cryopreservation Medium, MP Biomedicals
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
0920300M2
10220-624EA 213.92 USD
10220-624 10222-116 10222-118
Cellvation Cryopreservation Medium, MP Biomedicals
Cell Freezing Media
Cellvation is a unique cryopreservation medium designed for optimal cell recovery and convenience.
  • Optimal cell recovery and convenience
  • No DMSO or serum
  • Animal component free
  • Preserve stem cells, hepatocytes, and tissue samples.
Cellvation is a unique cryopreservation medium designed for optimal cell recovery and convenience. It does not contain DMSO or serum, and is scientifically engineered for Serum-Free cultures, as well as, ideally suited for all types of serum. Cellvation Cryopreservation Media is animal component free and has been shown to preserve the potential of stem cell for differentiation. This medium is recommended for the preservation of stem cells, hepatocytes, tissue samples, and a wide variety of cell types and established cell lines.
Osmomality: 1300 - 1600 mOsm/kg, Post Thaw Viability >75%