Kanamycin sulfate solution

Supplier: 0000012207

091672048
10221-874EA 107.95 USD
10221-874
Kanamycin sulfate solution
Kanamycin sulphate

Kanamycin is an aminoglycoside antibiotic effective against mycoplasma, gram-positive and gram-negative bacteria.


  • Effective against mycoplasma
  • Used in biotechnology applications
  • Inhibit protein synthesis


Kanamycin is an aminoglycoside antibiotic effective against mycoplasma, gram-positive and gram-negative bacteria


Mode of Action: Binds to 70S ribosomal subunit; inhibits translocation; elicits miscoding. Antimicrobial spectrum: Gram-negative and Gram-positive bacteria, and mycoplasma


It is used in biotechnology applications to inhibit protein synthesis.

Formula: C₁₈H₃₈N₄O₁₅S
MDL Number: MFCD00070253
CAS Number: 25389-94-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR