Fetalgro Bovine Growth Serum, RMBIO
Supplier: Rocky Mountain Biologicals
FGR-BBT
10221-866EA 262.41 USD
10221-866
Fetalgro Bovine Growth Serum, RMBIO
Sera (Serum)
Fetalgro® is Rocky Mountain Biologicals proprietary bovine growth serum (BGS) supplemented with additives that match or outperform FBS' cell growth capabilities in many cell lines.
Each lot of Fetalgro® is tested by RMBIO's team of experienced scientists in our own state-of-the-art cell culture lab prior to releasing the product for customer use. This investment in Fetalgro's product release testing gives us the ability to find the optimal performance for specific cell lines.
Certifications: Tested according to 9CFR 113.26, 28, 53