Anti-CD68 Rabbit Polyclonal Antibody

Supplier: Proteintech
25747-1-AP-150UL
10220-880EA 462.47 USD
10220-880
Anti-CD68 Rabbit Polyclonal Antibody
Antibodies
The CD68 antibody from Proteintech is a rabbit polyclonal antibody to a fusion protein of human CD68. This antibody recognizes human antigen. The CD68 antibody has been validated for the following applications: ELISA, IHC analysis.

Type: Primary
Antigen: CD68
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR