Anti-NFATC2 Rabbit Polyclonal Antibody

Supplier: Proteintech
22023-1-AP-150UL
10220-856EA 462.47 USD
10220-856
Anti-NFATC2 Rabbit Polyclonal Antibody
Antibodies
The NFATC2 antibody from Proteintech is a rabbit polyclonal antibody to a fusion protein of human NFATC2. This antibody recognizes human, mouse antigen. The NFATC2 antibody has been validated for the following applications: ELISA, WB, IF, IHC analysis.

Type: Primary
Antigen: NFATC2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR