Anti-CDX1 Rabbit Polyclonal Antibody

Supplier: Proteintech
21655-1-AP-150UL
10220-854EA 462.47 USD
10220-854
Anti-CDX1 Rabbit Polyclonal Antibody
Antibodies
The CDX1 antibody from Proteintech is a rabbit polyclonal antibody to a fusion protein of human CDX1. This antibody recognizes human, mouse antigen. The CDX1 antibody has been validated for the following applications: ELISA, WB analysis.

Type: Primary
Antigen: CDX1
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR