Anti-C14ORF156 Rabbit Polyclonal Antibody

Supplier: Proteintech
26006-1-AP-150UL
10220-822EA 462.47 USD
10220-822
Anti-C14ORF156 Rabbit Polyclonal Antibody
Antibodies
The C14orf156 antibody from Proteintech is a rabbit polyclonal antibody to a fusion protein of human C14orf156. This antibody recognizes human antigen. The C14orf156 antibody has been validated for the following applications: ELISA, WB analysis.

Type: Primary
Antigen: C14ORF156
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR