Anti-FOXC1 Rabbit Polyclonal Antibody

Supplier: Proteintech
55365-1-AP-150UL
10220-778EA 462.47 USD
10220-778
Anti-FOXC1 Rabbit Polyclonal Antibody
Antibodies
The FOXC1 antibody from Proteintech is a rabbit polyclonal antibody to a peptide of human FOXC1. This antibody recognizes human, mouse antigen. The FOXC1 antibody has been validated for the following applications: ELISA, WB analysis.

Type: Primary
Antigen: FOXC1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR