Penicillin : Streptomycin solution (10000 IU/mL, 10 mg/mL), sterile

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Danger

091670249
10220-718EA 40.37 USD
10220-718 76461-584
Penicillin : Streptomycin solution (10000 IU/mL, 10 mg/mL), sterile
Penicillin/Streptomycin

Double antibiotic solution containing 10,000 units/mL of penicillin and 10,000 µg/mL of streptomycin.


  • Dual antibiotic solution
  • Supplements cell culture media
  • Effective against most gram positive and gram negative bacteria


The antibiotics penicillin and streptomycin are used to prevent bacterial contamination of cell cultures due to their effective combined action against gram-positive and gram-negative bacteria. This solution contains 10,000 units/mL of penicillin and 10,000 µg/mL of streptomycin.


Penicillin-Streptomycin is used to supplement cell culture media to control bacterial contamination. It is effective against most gram positive and gram negative bacteria.


Caution: For laboratory use. Not for injection.

Storage Temperature: Freezer
CAS Number: 1406-05-9 (Penicillin) - 57-92-1 (Streptomycin)

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR