Penicillin : Streptomycin solution (5000 IU/mL, 5 mg/mL), sterile

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Danger

091670049
10220-716EA 34.06 USD
10220-716
Penicillin : Streptomycin solution (5000 IU/mL, 5 mg/mL), sterile
Penicillin/Streptomycin

Double antibiotic solution containing 5,000 units/mL of penicillin and 5,000 µg/mL of streptomycin.


  • Dual antibiotic solution
  • Supplements cell culture media
  • Effective against most gram positive and gram negative bacteria


The antibiotics penicillin and streptomycin are used to prevent bacterial contamination of cell cultures due to their effective combined action against gram-positive and gram-negative bacteria. This solution contains 5,000 units/mL of penicillin and 5,000 µg/mL of streptomycin.


Penicillin-Streptomycin is used to supplement cell culture media to control bacterial contamination. It is effective against most gram positive and gram negative bacteria.


Caution: For laboratory use. Not for injection.

Storage Temperature: Freezer
CAS Number: 1406-05-9 (Penicillin) - 57-92-1 (Streptomycin)

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR