Nalgene® Syringe Prefilter Plus, Glass Fiber + Cellulose Acetate, 25 mm, Thermo Scientific
Supplier: Thermo Fisher Scientific
722-2520
10218-468CS 381.25 USD
10218-468
Nalgene® Syringe Prefilter Plus, Glass Fiber + Cellulose Acetate, 25 mm, Thermo Scientific
Filters Syringe Filters
Conduct two-stage filtration for sterilization of highly viscous solutions or solutions with high particulate loads.
  • Sterile
  • Clear housing ring
For sterilization and clarification of aqueous biological fluids. Filter contains 0.2 µm cellulose acetate membrane with built-in glass fiber prefilter to provide two-stage filtration. Prefilter increases yield by preventing larger particles from clogging the downstream 0.2 µm CA membrane. Polypropylene housing with female Luer-Lok®inlet and male Luer slip outlet.
Packaging: Individually blister-packed.