Anti-MAD2L1 Rabbit Polyclonal Antibody

Supplier: Abgent
AP20713C
10218-446EA 589.62 USD
10218-446
Anti-MAD2L1 Rabbit Polyclonal Antibody
Antibodies
Western Blot: 1:1000

Type: Primary
Antigen: MAD2L1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR