Anti-DIAPH1 Rabbit Polyclonal Antibody

Supplier: Abgent
AP20655A
10218-274EA 589.62 USD
10218-274
Anti-DIAPH1 Rabbit Polyclonal Antibody
Antibodies
Western Blot: 1:500-1:1000,

Type: Primary
Antigen: DIAPH1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR