Anti-HSP90AA1 Rabbit Polyclonal Antibody

Supplier: Abgent
AP20646C
10218-242EA 589.62 USD
10218-242
Anti-HSP90AA1 Rabbit Polyclonal Antibody
Antibodies
Western Blot: 1:1000

Type: Primary
Antigen: HSP90AA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Center
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR