Anti-AGAP4 Rabbit Polyclonal Antibody

Supplier: Abgent
AP20603C
10218-234EA 589.62 USD
10218-234
Anti-AGAP4 Rabbit Polyclonal Antibody
Antibodies
Western Blot: 1:1000, Immunhistochemsitry: 1:25, Flow Cytometry: 1:25, Immunofluorescence: 1:25,

Type: Primary
Antigen: AGAP4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR