Anti-UIMC1 Rabbit Polyclonal Antibody

Supplier: Abgent
AP20602C
10218-230EA 589.62 USD
10218-230
Anti-UIMC1 Rabbit Polyclonal Antibody
Antibodies
Western Blot: 1:1000, Flow Cytometry: 1:25,

Type: Primary
Antigen: UIMC1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR