Anti-HIST1H4A Rabbit Polyclonal Antibody

Supplier: Abgent
AP20585C
10218-222EA 589.3 USD
10218-222
Anti-HIST1H4A Rabbit Polyclonal Antibody
Antibodies
Western Blot: 1:2000, Immunhistochemsitry: 1:25, Flow Cytometry: 1:25,

Type: Primary
Antigen: HIST1H4A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR