Anti-PLAUR Rabbit Polyclonal Antibody

Supplier: Abgent
AP20579C
10218-218EA 561.55 USD
10218-218
Anti-PLAUR Rabbit Polyclonal Antibody
Antibodies
Immunhistochemsitry: 1:25,

Type: Primary
Antigen: PLAUR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR