Anti-HNF1BB Rabbit Polyclonal Antibody

Supplier: Abgent
AZB18694A
10218-134EA 589.62 USD
10218-134
Anti-HNF1BB Rabbit Polyclonal Antibody
Antibodies
Western Blot: 1:1000

Type: Primary
Antigen: HNF1BB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Center
Host: Rabbit
Isotype:
Reactivity: Zebrafish
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR