Anti-CDA Mouse Monoclonal Antibody [clone: 921CT14.2.1]

Supplier: Abgent
AM2218B
10217-956EA 586.14 USD
10217-956
Anti-CDA Mouse Monoclonal Antibody [clone: 921CT14.2.1]
Antibodies
Western Blot: 1:1000

Type: Primary
Antigen: CDA
Clonality: Monoclonal
Clone: 921CT14.2.1
Conjugation: Unconjugated
Epitope: Center
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR