Anti-JUN Mouse Monoclonal Antibody [clone: 1306CT545.208.117]

Supplier: Abgent
AM8428B
10217-904EA 589.62 USD
10217-904
Anti-JUN Mouse Monoclonal Antibody [clone: 1306CT545.208.117]
Antibodies
Western Blot: 1:1000

Type: Primary
Antigen: JUN
Clonality: Monoclonal
Clone: 1306CT545.208.117
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR