Anti-USF1 Mouse Monoclonal Antibody [clone: 1264CT170.274.14]

Supplier: Abgent
AM2248B
10217-880EA 586.14 USD
10217-880
Anti-USF1 Mouse Monoclonal Antibody [clone: 1264CT170.274.14]
Antibodies
Western Blot: 1:500,

Type: Primary
Antigen: USF1
Clonality: Monoclonal
Clone: 1264CT170.274.14
Conjugation: Unconjugated
Epitope: Center
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR