Anti-HDAC2 Mouse Monoclonal Antibody [clone: 1194CT18.5.1]

Supplier: Abgent
AM2246B
10217-874EA 558.24 USD
10217-874
Anti-HDAC2 Mouse Monoclonal Antibody [clone: 1194CT18.5.1]
Antibodies
Western Blot: 1:1000

Type: Primary
Antigen: HDAC2
Clonality: Monoclonal
Clone: 1194CT18.5.1
Conjugation: Unconjugated
Epitope: Center
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR