Anti-CREB3L4 Mouse Monoclonal Antibody [clone: 1165CT16.1.1]

Supplier: Abgent
AM2245B
10217-868EA 589.62 USD
10217-868
Anti-CREB3L4 Mouse Monoclonal Antibody [clone: 1165CT16.1.1]
Antibodies
Western Blot: 1:1000

Type: Primary
Antigen: CREB3L4
Clonality: Monoclonal
Clone: 1165CT16.1.1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR