Anti-ERBB2 Rabbit Polyclonal Antibody

Supplier: Abgent
AP3900A
10217-862EA 561.55 USD
10217-862
Anti-ERBB2 Rabbit Polyclonal Antibody
Antibodies
Western Blot: 1:4000,

Type: Primary
Antigen: ERBB2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR